Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ZNF596 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563131
Description
Immunogen sequence: LVSIGKQLCK SVVLSQLEQV EKLSTQRISL LQGREVGIKH QEIPFIQHIY QKGTSTISTM RSHTQEDP Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 30%.
May be involved in transcriptional regulation.
            Specifications
| ZNF596 | |
| Polyclonal | |
| Unconjugated | |
| ZNF596 | |
| Zinc finger protein 596; ZNF596 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 169270 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | 
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC21 | |
| ZNF596 | |
| Recombinant protein corresponding to Human ZNF596. Recombinant protein control fragment (Product #RP-101487). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction