Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF598 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559712
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559712 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559712 Supplier Invitrogen™ Supplier No. PA559712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVGGEDYEEV DRYSRQGRVA RAGTRGAQQS RRGSWRYKRE EEDREVAAAV RASVAAQQQE EARRSEDQEE GGRPKKEEAA ARGPEDPRGP RRSPRTQGEG PGPKETSTNG PVSQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.

ZNF598 (E3 ubiquitin-protein ligase ZNF598) plays a key role in the ribosome quality control (RQC). RQC is a pathway that takes place when a ribosome has stalled during translation. It is required for ribosomes to terminally stall during translation of poly(A) sequences by mediating monoubiquitination of 40A ribosomal protein RPS10/eS10 and RPS3/uS3. Stalling precludes synthesis of a long poly-lysine tail and initiated the RQC pathway to degrade the potentially detrimental aberrant nascent polypeptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF598
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF598
Gene Accession No. Q86UK7
Gene Alias BC023040; C6orf149; E3 ubiquitin-protein ligase ZNF598; FLJ00086; ISD11; Ntrap; Zfp598; Zinc finger protein 598; ZNF598
Gene Symbols ZNF598
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF598. Recombinant protein control fragment (Product #RP-98748).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90850
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.