Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF599 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569688
Description
This target displays homology in the following species: Cow: 92%; Human: 100%.
May be involved in transcriptional regulation.
Specifications
ZNF599 | |
Polyclonal | |
Unconjugated | |
ZNF599 | |
Zinc finger protein 599; ZNF599 | |
Rabbit | |
Affinity chromatography | |
RUO | |
148103 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96NL3 | |
ZNF599 | |
synthetic peptide directed towards the N terminal region of human ZNF599. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction