Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF608 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552160 Supplier Invitrogen™ Supplier No. PA552160
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPGTPKGKRE LMSNGPGSII GAKAGKNSGK KKGLNNELNN LPVISNMTAA LDSCSAADGS LAAEMPKLEA EGLIDKKNLG DKEKGKKATN CKTDKNLSKL KSARPIAPAP APTPPQLIAI PTATFTTTTT GTIPGLPSLT TTVVQATP Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

ZNF608 gene ontology annotations related to this gene include nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF608
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF608
Gene Accession No. Q9ULD9
Gene Alias 4932417D18Rik; D430007A19Rik; Kiaa1281; LOW QUALITY PROTEIN: zinc finger protein 608; NY-REN-36; renal carcinoma antigen NY-REN-36; Zfp608; Zinc finger protein 608; Znf608
Gene Symbols ZNF608
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF608. Recombinant protein control fragment (Product #RP-101065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57507
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.