Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF609 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559276
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559276 Supplier Invitrogen™ Supplier No. PA559276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEGKVDSVK SKDAEQLVKE GAKKTLFPPQ PQSKDSPYYQ GFESYYSPSY AQSSPGALNP SSQAGVESQA LKTKRDEEPE SIEGKVKNDI CEEKKPE Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.

The ZNF609 gene encodes zinc finger protein 609, which functions as a transcriptional regulator involved in the modulation of gene expression during development and cellular differentiation. ZNF609 contains zinc finger domains, which enable it to bind DNA and control the transcription of target genes, influencing processes such as cell proliferation and neuronal development. This gene is particularly important for normal neural and muscle cell function and has been implicated in neurodevelopmental disorders and cancer. ZNF609 is known to interact with signaling pathways that control neuron differentiation and migration, and alterations in its expression have been associated with conditions like intellectual disability and developmental delay. Furthermore, ZNF609's role in cancer biology shows its involvement in regulating genes linked to tumor progression and metastasis. Studies on ZNF609 focus on understanding its regulatory mechanisms and potential as a therapeutic target for diseases related to developmental deficits and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF609
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF609
Gene Accession No. O15014
Gene Alias 9830165N24; BC016271; Kiaa0295; mKIAA0295; RGD1561651; Zfp609; Zinc finger protein 609; zinc finger protein 609 pseudogene; Znf609
Gene Symbols ZNF609
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF609. Recombinant protein control fragment (Product #RP-98150).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23060
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.