Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF610 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598265
Description
ZNF610 Polyclonal Antibody for Western Blot, ELISA
May be involved in transcriptional regulation.
Specifications
ZNF610 | |
Polyclonal | |
Unconjugated | |
ZNF610 | |
FLJ36040; Zinc finger protein 610; zink finger protein; ZNF610 | |
Rabbit | |
Protein G | |
RUO | |
162963 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8N9Z0 | |
ZNF610 | |
Recombinant Human Zinc finger protein 610 protein (1-200AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction