Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF611 Monoclonal Antibody (4F1)
GREENER_CHOICE

Catalog No. PIMA522687
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522687 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522687 Supplier Invitrogen™ Supplier No. MA522687
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MLQRLQIIGE SIMKRDLTSV INVANFSEIV HILQVIGELI LERNLTNVMT VPRSSVKLHP MQNIGEFIQE RNLTSVMIVA KPLLHVHTSL DIRESILDRN LTNVISVARS SVQDHSMQNI RKFIFEITVP NEMSIANHQA LIDIGVNSAL T.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF611
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4F1
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF611
Gene Accession No. Q8N823
Gene Alias C6orf55; Zinc finger protein 611; ZNF611
Gene Symbols ZNF611
Host Species Mouse
Immunogen ZNF611 (AAH00918, 1 a.a. ~ 151 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81856
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.