Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF614 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112806
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112806 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112806 Supplier Invitrogen™ Supplier No. PA5112806
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF614 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF614 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF614
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF614
Gene Accession No. Q8N883
Gene Alias LOW QUALITY PROTEIN: zinc finger protein 614; zinc finger protein 614; ZNF614
Gene Symbols ZNF614
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 614 protein (1-198AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80110
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.