Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF620 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116668 Supplier Invitrogen™ Supplier No. PA5116668
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cellular location: Nucleus Immunogen sequence: MVGGLPGNVS QHLDFGSSLE QPQGHWIIKT KSKRRHFTDT SARHHEAYEV KNGEKFEKLG KNISVSTQLT TNQTNPSGQI SYECGQCGRY FIQMADFHRH EKCHTGEKSF ECKECGKYFR YNSLLIRHQI IHTGKKPFKC KECGKGLSSD.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF620
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF620
Gene Accession No. Q6ZNG0
Gene Alias Zinc finger protein 620; ZNF620
Gene Symbols ZNF620
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human ZNF620 (NP_0012430961).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253639
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.