Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF622 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232087JF549
Description
ZNF622 Polyclonal antibody specifically detects ZNF622 in Human, Mouse samples. It is validated for Western BlotSpecifications
ZNF622 | |
Polyclonal | |
Western Blot | |
MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Zinc Finger Protein 622 using the numbering given in entry NP_219482.1 (GeneID 90441). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
90441 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction