Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF622 Antibody (PCRP-ZNF622-1C11), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308811PE
Description
ZNF622 Monoclonal antibody specifically detects ZNF622 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
ZNF622 | |
Monoclonal | |
PE | |
MGC2485, zinc finger protein 622, Zinc finger-like protein 9, ZPR9MGC17552 | |
Recombinant full-length human ZNF622 protein (Uniprot: Q969S3) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-ZNF622-1C11 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
90441 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction