Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF625 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567321
Description
Immunogen sequence: HTGEKPCSSNT SKGQGEKIA Highest antigen sequence identity to the following orthologs - mouse 55%, rat 55%.
ZNF625 may be involved in transcriptional regulation.
Specifications
ZNF625 | |
Polyclonal | |
Unconjugated | |
ZNF625 | |
Zinc finger protein 625; ZNF625 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90589 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96I27 | |
ZNF625 | |
Recombinant Human ZNF625. Recombinant protein control fragment (Product #RP-107964). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction