Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF628 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569718 Supplier Invitrogen™ Supplier No. PA569718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 79%; Rat: 79%.

Zinc finger proteins (ZNFs), which bind nucleic acids, perform many key functions, the most important of which is regulating transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF628
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF628
Gene Accession No. Q5EBL2
Gene Alias Zec; Zfp628; Zinc finger protein 628; zinc finger protein expressed in embryonal cells and certain adult organs; zinc finger protein Zec; Znf628
Gene Symbols ZNF628
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ZNF628.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89887
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.