Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF630 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564875 Supplier Invitrogen™ Supplier No. PA564875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VTFEDVAVDFT QEEWQQLNPA QKTLHRDVML ETYNHLVSVG CSGIKPDVIF KLEHGKDPWI IESELSRWIY PDRVKGLESS QQIISGELLF QREILERAPK DNSL Highest antigen sequence identity to the following orthologs - mouse 50%, rat 46%.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF630
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF630
Gene Accession No. Q2M218
Gene Alias dJ54B20.2; dJ54B20.2 (novel KRAB box containing C2H2 type zinc finger protein); zinc finger protein 630; ZNF630
Gene Symbols ZNF630
Host Species Rabbit
Immunogen Recombinant Human ZNF630. Recombinant protein control fragment (Product #RP-105680).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.