Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF630 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566979
Description
Immunogen sequence: SQQIISGELLF QREILERAPK DNSLYSVLKI WHIDNQMDRY QGNQDRVLRQ VTVISRETLT DEMGSKYSAF GKMFNRCTDL APLSQKFHKF DSCE Highest antigen sequence identity to the following orthologs - mouse 29%, rat 28%.
May be involved in transcriptional regulation.
Specifications
ZNF630 | |
Polyclonal | |
Unconjugated | |
ZNF630 | |
dJ54B20.2; dJ54B20.2 (novel KRAB box containing C2H2 type zinc finger protein); zinc finger protein 630; ZNF630 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57232 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M218 | |
ZNF630 | |
Recombinant Human ZNF630. Recombinant protein control fragment (Product #RP-107623). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction