Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF638/NP220 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560667
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0667 Supplier Bethyl Laboratories, Inc Supplier No. A301548A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.

Specifications

Antigen ZNF638/NP220
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZNF638
Gene Accession No. Q14966
Gene Alias Zinc finger protein 638, Cutaneous T-cell lymphoma-associated antigen se33-1, CTCL-associated antigen se33-1, Nuclear protein 220, Zinc finger matrin-like protein, NP220, ZFML
Gene Symbols ZNF638
Host Species Rabbit
Immunogen Between 1928 and 1978
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27332
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.