Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF638 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537001
Description
This antibody detects endogenous protein at a molecular weight of 220 kDa and 120 kDa (Isoform 5). Purity is >95% by SDS-PAGE.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.
Specifications
ZNF638 | |
Polyclonal | |
Unconjugated | |
ZNF638 | |
AI323587; CTCL tumor antigen se33-1; CTCL-associated antigen se33-1; Cutaneous T-cell lymphoma-associated antigen se33-1; mNP220; Np220; NP220 nuclear protein; nuclear protein 220; Zfml; Zfp638; Zinc finger matrin-like protein; zinc finger protein 638; zinc finger, matrin-like; Znf638 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18139, 27332, 312491 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14966, Q61464 | |
Zfp638, ZNF638 | |
Synthetic peptide corresponding to amino acids 1308-1353 of human ZNF638. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction