Learn More
Invitrogen™ ZNF638 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590734
Description
Immunogen sequence: MSRPRFNPRG DFPLQRPRAP NPSGMRPPGP FMRPGSMGLP RFYPAGRARG IPHRFAGHES YQNMGPQRMN VQVTQHRTDP RLTKEKLDFH EAQQKKGKPH GSRWDDEPHI SASVAVKQSS VTQVTEQSPK VQSRYTKESA SSILASFGLS NEDLEELSRY PDEQLTPENM PLILRDIRMR KMGRRLPNLP SQSRNKETLG SEAVSSNVID YGHASKYGYT EDPLEVRIYD PEIPTDEVEN EFQSQQNISA SVPNPNVICN SMFPVEDVFR QMDFPGESSN NRSFFSVESG TKMSGLHISG; Positive Samples: Mouse brain; Cellular Location: Nucleus speckle.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. NP220 (Nuclear protein 220), also designated Zinc finger protein 638 (ZNF638) or Cutaneous T-cell lymphoma-associated antigen se33-1, is a 1978 amino acid protein that contains one matrin-type zinc finger and two RRM (RNA recognition motif) domains, suggesting a role in transcriptional regulation. NP220 binds to double-stranded DNA fragments by recognizing clusters of cytidines. NP220 interacts with FHL-2 and is also thought to be phosphorylated by ATM or ATR upon DNA damage. It exists as five isoforms as a result of alternative splicing events. Isoform five of NP220 is a tumor-associated antigen found in several cutaneous T-cell lymphoma (CTCL), and in particular in mycosis fungoides patients and in Sezary syndrome patients.
Specifications
ZNF638 | |
Polyclonal | |
Unconjugated | |
ZNF638 | |
AI323587; CTCL tumor antigen se33-1; CTCL-associated antigen se33-1; Cutaneous T-cell lymphoma-associated antigen se33-1; mNP220; Np220; NP220 nuclear protein; nuclear protein 220; Zfml; Zfp638; Zinc finger matrin-like protein; zinc finger protein 638; zinc finger, matrin-like; Znf638 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18139, 27332 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.43 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q14966, Q61464 | |
Zfp638, ZNF638 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZNF638 (NP_0012395421). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.