Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF639 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561864
Description
Immunogen sequence: SADIVICDEE CDSPESVNQQ TQEESPIEVH TAEDVPIAVE VHAISEDYDI ETENNSSESL QDQTDEEPPA KLCKILDKSQ ALNVTAQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
Specifications
ZNF639 | |
Polyclonal | |
Unconjugated | |
ZNF639 | |
6230400O18Rik; ANC_2H01; ANC-2H01; ZASC1; Zfp639; zinc finger amplified in esophageal squamous cell carcinomas 1; zinc finger protein 639; Zinc finger protein 639-like protein; Zinc finger protein ANC_2H01; Zinc finger protein ZASC1; ZNF639 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51193 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UID6 | |
ZNF639 | |
Recombinant protein corresponding to Human ZNF639. Recombinant protein control fragment (Product #RP-91624). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction