Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF639 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562564 Supplier Invitrogen™ Supplier No. PA562564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRKTLHPSRY SDSSGISRIA DGFNGIFSDH CYSVCSMRQP DLKYFDNKDD DSDTETSNDL PKFADGIKAR NRNQNYLVPS PVL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 88%.

This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF639
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF639
Gene Accession No. Q9UID6
Gene Alias 6230400O18Rik; ANC_2H01; ANC-2H01; ZASC1; Zfp639; zinc finger amplified in esophageal squamous cell carcinomas 1; zinc finger protein 639; Zinc finger protein 639-like protein; Zinc finger protein ANC_2H01; Zinc finger protein ZASC1; ZNF639
Gene Symbols ZNF639
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF639. Recombinant protein control fragment (Product #RP-91623).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51193
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.