Learn More
ZNF639 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302400AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
ZNF639 binds DNA and may function as a transcriptional repressor.Specifications
ZNF639 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UID6 | |
ZNF639 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ZNF639 | |
6230400O18Rik, ANC-2H01, ANC_2H01, ZASC1, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein ANC_2H01, zinc finger protein ZASC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51193 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.