Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF639 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501562493
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-2493 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-2493 Supplier Bethyl Laboratories, Inc Supplier No. A302401AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine

ZNF639 binds DNA and may function as a transcriptional repressor.

Specifications

Antigen ZNF639
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ZNF639
Gene Accession No. Q9UID6
Gene Alias 6230400O18Rik, ANC-2H01, ANC_2H01, ZASC1, zinc finger amplified in esophageal squamous cell carcinomas 1, zinc finger protein ANC_2H01, zinc finger protein ZASC1
Gene Symbols ZNF639
Host Species Rabbit
Immunogen Between 435 and 485
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51193
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.