Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF643 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560667
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560667 Supplier Invitrogen™ Supplier No. PA560667
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KTKESALQND ISWEELHCGL MMERFTKGSS MYSTLGRISK CNKLESQQEN QRMGKGQIPL MCKKTFTQER GQESNRFEKR INVKSEVMPG PIGLPRKRDR KYDTPGKRSR YNIDLVNHSR SYT Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 29%.

May be involved in transcriptional regulation. Essential for Golgi structural integrity (PubMed:29851555). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF643
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZFP69B
Gene Accession No. Q9UJL9
Gene Alias novel zinc finger protein; ZFP69 zinc finger protein B; ZFP69B; Zinc finger protein 643; Zinc finger protein 69 homolog B; zinc finger protein ZFP69B; ZKSCAN23B; ZNF643; ZSCAN54B
Gene Symbols ZFP69B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF643. Recombinant protein control fragment (Product #RP-91315).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.