Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF644 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178195AF488
Description
ZNF644 Polyclonal antibody specifically detects ZNF644 in Human samples. It is validated for Western BlotSpecifications
ZNF644 | |
Polyclonal | |
Western Blot | |
BM-005, KIAA1221Zep-2, MGC60165, MGC70410, ZEP2, zinc finger motif enhancer binding protein 2, Zinc finger motif enhancer-binding protein 2, zinc finger protein 644 | |
The immunogen for this product maps to a region between residue 1 and 50 of human Zinc Finger Protein 644 using the numbering given in entry NP_958357.1 (GeneID 84146). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
84146 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction