Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF644 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563487
Description
Immunogen sequence: NGPVSHSSLT KTSNMNKGSV SLTTGQPVDQ PTTESCSTLK VAADLQLSTP QKASQHQVLF LLSDVAHAKN PTHSNKKLPT SASVGCDIQN SVGSNIKS Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 69%.
May be involved in transcriptional regulation.
Specifications
ZNF644 | |
Polyclonal | |
Unconjugated | |
ZNF644 | |
BM-005; KIAA1221; MGC60165; MGC70410; MYP21; NatF; ZEP2; Zep-2; zinc finger motif enhancer binding protein 2; Zinc finger motif enhancer-binding protein 2; Zinc finger protein 644; Zinc finger protein 644-like protein; ZNF644 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84146 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H582 | |
ZNF644 | |
Recombinant protein corresponding to Human ZNF644. Recombinant protein control fragment (Product #RP-101245). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction