Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF645 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531534
Description
Recommended positive controls: MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZNF645 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions.
Specifications
| ZNF645 | |
| Polyclonal | |
| Unconjugated | |
| ZNF645 | |
| Cbl proto-oncogene-like protein 2; CBLL2; c-Cbl-like protein 2; CT138; E3 ubiquitin ligase; E3 ubiquitin-protein ligase CBLL2; E3 ubiquitin-protein ligase ZNF645; HAKAIL; RING-type E3 ubiquitin transferase ZNF645; Zinc finger protein 645; ZNF645 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8N7E2 | |
| ZNF645 | |
| Recombinant protein encompassing a sequence within the center region of human ZNF645. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction