Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF645 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598669
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598669 Supplier Invitrogen™ Supplier No. PA598669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF645 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

ZNF645 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF645
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF645
Gene Accession No. Q8N7E2
Gene Alias Cbl proto-oncogene-like protein 2; CBLL2; c-Cbl-like protein 2; CT138; E3 ubiquitin ligase; E3 ubiquitin-protein ligase CBLL2; E3 ubiquitin-protein ligase ZNF645; HAKAIL; RING-type E3 ubiquitin transferase ZNF645; Zinc finger protein 645; ZNF645
Gene Symbols ZNF645
Host Species Rabbit
Immunogen Recombinant Human E3 ubiquitin-protein ligase CBLL2 protein (200-413AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158506
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.