Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF652 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540418
Description
Peptide sequence: RENSDDTEEE EEEVSYKREQ IIVEVNLNNQ TLNVSKGEKG VSSQSKETPV Sequence homology: Cow: 88%; Dog: 93%; Goat: 91%; Horse: 87%; Human: 100%; Mouse: 93%; Pig: 78%; Rabbit: 90%; Rat: 93%; Yeast: 91%.
ZNF652 is a new candidate transcription factor.
Specifications
| ZNF652 | |
| Polyclonal | |
| Unconjugated | |
| ZNF652 | |
| 9530033F24Rik; DKFZp781E2122; KIAA0924; RGD1566329; Zfp652; Zinc finger protein 652; Znf652 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y2D9 | |
| ZNF652 | |
| Synthetic peptide directed towards the N-terminal of human ZNF652 (aa 97-146). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction