Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF652 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116875 Supplier Invitrogen™ Supplier No. PA5116875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Jurkat, BxPC-3, Mouse liver, Rat brain Immunogen sequence: MSHTASSCQE LVENCAVHVA GMAQEDSRRG QVPSSFYHGA NQELDLSTKV YKRESGSPYS VLVDTKMSKP HLHE.

ZNF652 is a new candidate transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF652
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZNF652
Gene Accession No. A1L1J6, Q5DU09, Q9Y2D9
Gene Alias 9530033F24Rik; DKFZp781E2122; KIAA0924; RGD1566329; Zfp652; Zinc finger protein 652; Znf652
Gene Symbols Zfp652, ZNF652
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-74 of human ZNF652 (NP_0557121).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22834, 268469, 497984
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.