Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF653 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554854 Supplier Invitrogen™ Supplier No. PA554854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NVNCLKNVVI WYEDHKHRCP YEPHLAELDP TFGLYTTAVW QCEAGHRYFQ DLHSPLKPLS DSDPDSDKVG NGLVAGSSDS SSSGS Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF653
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF653
Gene Accession No. Q96CK0
Gene Alias 67 kDa zinc finger protein; E430039K05Rik; zinc finger protein 653; Zinc finger protein Zip67; ZIP67; ZNF653
Gene Symbols ZNF653
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF653. Recombinant protein control fragment (Product #RP-93599).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115950
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.