Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF654 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232110B
Description
ZNF654 Polyclonal antibody specifically detects ZNF654 in Human samples. It is validated for Western BlotSpecifications
ZNF654 | |
Polyclonal | |
Western Blot | |
FLJ10997, FLJ21142, zinc finger protein 654 | |
The immunogen this antibody was made to, maps to a region between residue 477 to 527 of human Zinc Finger Protein 654 using the numbering given in entry NP_060763.2 (GeneID 55279). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55279 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction