Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF655 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178732AF594
Description
ZNF655 Polyclonal antibody specifically detects ZNF655 in Human samples. It is validated for Western BlotSpecifications
ZNF655 | |
Polyclonal | |
Western Blot | |
zinc finger protein 655 | |
The immunogen for this product maps to a region between residue 1 and 50 of human Vav-1 Interacting Kruppel-like Protein using the numbering given in entry NP_612503.1 (GeneID 79027). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
79027 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction