Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF660 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541244
Description
Peptide sequence: TFRQTSQVIL HLRTHTKEKP YKCSECGKAY RYSSQLIQHQ RKHNEEKETS Sequence homology: Horse: 100%; Human: 100%; Rabbit: 86%; Rat: 93%.
Located on chromosome 3, the ZNF660 gene encodes for zinc finger protein 660.
Specifications
| ZNF660 | |
| Polyclonal | |
| Unconjugated | |
| ZNF660 | |
| Zinc finger protein 660; ZNF660 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 285349 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6AZW8 | |
| ZNF660 | |
| Synthetic peptide directed towards the C-terminal of human ZNF660 (aa 282-331). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction