Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF662 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ZNF662 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZNF662 Polyclonal specifically detects ZNF662 in Human samples. It is validated for Western Blot.Specifications
ZNF662 | |
Polyclonal | |
Rabbit | |
NP_001128128 | |
389114 | |
Synthetic peptide directed towards the middle region of human ZNF662The immunogen for this antibody is ZNF662. Peptide sequence QIFYICEECGKCFDQNEDFDQHQKTHNGEKVYGCKECGKAFSFRSHCIAH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ33347, zinc finger protein 662 | |
ZNF662 | |
IgG | |
51 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title