Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF664 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569782
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
May be involved in transcriptional regulation.
Specifications
ZNF664 | |
Polyclonal | |
Unconjugated | |
ZNF664 | |
AW060232; D930038J03Rik; ZFOC1; Zfp664; zinc finger Organ of Corti 1; zinc finger protein; zinc finger protein 176; Zinc finger protein 664; zinc finger protein from organ of Corti; zinc finger protein ZFOC1; ZNF176; ZNF196; Znf664 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
144348 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N3J9 | |
ZNF664 | |
synthetic peptide directed towards the N terminal region of human ZNF664. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction