Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF665 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541177
Description
Peptide sequence: CGKAFTVRSN LTIHQVIHTG EKPYKCNECG KVFSQPSNLA GHQRIHTGEK Sequence homology: Cow: 79%; Dog: 79%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Mouse: 86%; Rabbit: 85%; Rat: 86%.
ZNF665 is a new candidate transcription factor. Western blots using two different antibodies (ARP36042-T200 and ARP36043_T200) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications
ZNF665 | |
Polyclonal | |
Unconjugated | |
ZNF665 | |
ZFP160L; Zinc finger protein 160-like; Zinc finger protein 665; ZNF665 | |
Rabbit | |
Protein A | |
RUO | |
79788 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H7R5 | |
ZNF665 | |
Synthetic peptide directed towards the N-terminal of human ZNF665 (aa 211-260). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction