Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF665 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541176 Supplier Invitrogen™ Supplier No. PA541176
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LGVSFQSHLP ELQQFQREGK IYEYNQVEKS PNNRGKHYKC DECGKVFSQN Sequence homology: Dog: 85%; Horse: 91%; Human: 100%; Pig: 82%; Rabbit: 90%.

ZNF665 is a new candidate transcription factor. Western blots using two different antibodies (ARP36042-T200 and ARP36043_T200) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF665
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF665
Gene Accession No. Q9H7R5
Gene Alias ZFP160L; Zinc finger protein 160-like; Zinc finger protein 665; ZNF665
Gene Symbols ZNF665
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF665.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79788
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.