Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF667 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561278 Supplier Invitrogen™ Supplier No. PA561278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APWMVEPVRR RRAPDSGSKC ETKKLPPNQC NKSGQSICQK LVSAQQKAPT RKSGCNKNSV LVKPKKGHSG K Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 70%.

Mipu1, also known as zinc finger protein 667 or ZNF667, encodes a nuclear-localized protein containing 14 carboxy-terminal zinc finger motifs and an amino-terminal KRAB domain. This protein is highly expressed in heart and brain and is upregulated in rat heart after a transient ischemia-reperfusion procedure. Overexpression experiments suggest that Mipu1 suppresses the transcriptional activities of AP-1 and SRE in the MAPK signaling pathway and thus may play a role in the pathogenesis of cardiac and vascular disease. At least four isoforms of MIPU1 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF667
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF667
Gene Accession No. Q5HYK9
Gene Alias A830025F02Rik; Mip1; MIPU1; myocardial ischemic preconditioning upregulated 1; myocardial ischemic preconditioning upregulated 1 ortholog; Myocardial ischemic preconditioning up-regulated protein 1; Zfp667; Zinc finger protein 667; Znf667
Gene Symbols ZNF667
Host Species Rabbit
Immunogen Recombinant protein fragment of Human ZNF667 (Product #RP-95766)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63934
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.