Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF671 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ZNF671 |
---|---|
Concentration | 0.71 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3373
![]() ![]() |
Proteintech Group Inc
213291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ZNF671 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 10 C2H2-type zinc fingers and 1 KRAB domain. ZNF671 may be involved in transcriptional regulation.Specifications
ZNF671 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q8TAW3 | |
79891 | |
ZNF671 Fusion Protein Ag15319 | |
Primary | |
-20°C | |
ZNF671 |
0.71 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
zinc finger protein 671, ZNF671 | |
ZNF671 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title