Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Znf672 Polyclonal Antibody

Catalog No. PIPA532150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532150 Supplier Invitrogen™ Supplier No. PA532150

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

May be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Znf672
Applications Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene ZNF672
Gene Accession No. Q499Z4, Q99LH4
Gene Alias 4930488P06Rik; 4930511N19Rik; FLJ22301; Zfp672; Zinc finger protein 672; ZN672; Znf672
Gene Symbols Zfp672, ZNF672
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF672. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319475, 79894
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.