Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF675 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116272
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116272 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116272 Supplier Invitrogen™ Supplier No. PA5116272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZNF675 Polyclonal Antibody for Western Blot

ZNF675 may be involved in transcriptional regulation. It may play a role during osteoclast differentiation by modulating TRAF6 signaling activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF675
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ZNF675
Gene Accession No. Q8TD23
Gene Alias TBZF; TIZ; TRAF6 inhibitory zinc finger; TRAF6-binding zinc finger protein; TRAF6-inhibitory zinc finger protein; zinc finger protein 675; ZNF675
Gene Symbols ZNF675
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 136-162 amino acids from the N-terminal region of human ZNF675.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.