Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF677 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590163 Supplier Invitrogen™ Supplier No. PA590163
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNIPPEDDIS VGFTSKGLSP KENNKEELYH LVILERKESH GINNFDLKEV WENMPKFDSL WDYDVKNYKG MPLTCNKNLT HRKDQQHNKS SIHFSLKQSV SIRDSAHQYF IHDKPFIRNL LKLKNNIRYA GNKYVKCFEN KIGLSLQAQL A; Positive Samples: Rat testis; Cellular Location: Nucleus.

ZNF677 (Zinc Finger Protein 677) is a Protein Coding gene. Among its related pathways areGene Expression. Gene Ontology (GO) annotations related to this gene includenucleic acid bindingandDNA-binding transcription factor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF677
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias hypothetical protein MGC48625; zinc finger protein 677; ZNF677
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human ZNF677 (NP_8724151).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.