Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF677 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110607
Description
Immunogen sequence: DNIPPEDDIS VGFTSKGLSP KENNKEELYH LVILERKESH GINNFDLKEV WENMPKFDSL WDYDVKNYKG MPLTCNKNLT HRKDQQHNKS SIHFSLKQSV SIRDSAHQYF IHDKPFIRNL LKLKNNIRYA GNKYVKCFEN KIGLSLQAQL A.
ZNF677 (Zinc Finger Protein 677) is a Protein Coding gene. Among its related pathways areGene Expression. Gene Ontology (GO) annotations related to this gene includenucleic acid bindingandDNA-binding transcription factor activity.
Specifications
ZNF677 | |
Polyclonal | |
Unconjugated | |
ZNF677 | |
hypothetical protein MGC48625; zinc finger protein 677; ZNF677 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
342926 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.47 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q86XU0 | |
ZNF677 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human ZNF677 (NP_872415.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction