Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF677 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554697
Description
Immunogen sequence: NYRNLLSLDE DNIPPEDDIS VGFTSKGLSP KENNKEELYH LVILERKESH GINNFDLKEV WENMPKFDSL WDYDVK Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
ZNF677 (Zinc Finger Protein 677) is a Protein Coding gene. Among its related pathways areGene Expression. Gene Ontology (GO) annotations related to this gene includenucleic acid bindingandDNA-binding transcription factor activity.
Specifications
ZNF677 | |
Polyclonal | |
Unconjugated | |
ZNF677 | |
hypothetical protein MGC48625; zinc finger protein 677; ZNF677 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
342926 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86XU0 | |
ZNF677 | |
Recombinant protein corresponding to Human ZNF677. Recombinant protein control fragment (Product #RP-93648). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction