Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF679 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543655 Supplier Invitrogen™ Supplier No. PA543655
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VQKGGCNEVN QCLSTTQNKI FQTHKCVKVF GKFSNSNRHK TRHTGKKHFK Sequence homology: Human: 100%.

ZNF679 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF679
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF679
Gene Accession No. Q8IYX0
Gene Alias hypothetical protein MGC42415; zinc finger protein 679; ZNF679
Gene Symbols ZNF679
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF679 (aa 136-185).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168417
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.