Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF681 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543711
Description
Peptide sequence: PWTRKRHRMV AEPPVICSHF AQDFSPEQNI KDSFQKVTPR RYGKCEHENL Sequence homology: Human: 100%.
ZNF681 belongs to the krueppel C2H2-type zinc-finger protein family. It Contains 16 C2H2-type zinc fingers. ZNF681 may be involved in transcriptional regulation.
Specifications
ZNF681 | |
Polyclonal | |
Unconjugated | |
ZNF681 | |
Zinc finger protein 681; ZNF681 | |
Rabbit | |
Affinity chromatography | |
RUO | |
148213 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96N22 | |
ZNF681 | |
Synthetic peptide directed towards the N-terminal of human ZNF681 (aa 62-111). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction