Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF683 Mouse anti-Human, Polyclonal, Abnova™

Catalog No. 89339560 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-560 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-560 Supplier Abnova Corporation Supplier No. H00257101B01P
Add to Cart
Add to Cart

Mouse Polyclonal Antibody.

ZNF683 Polyclonal antibody specifically detects ZNF683 in Human samples. It is validated for IF, WB-Tr.

Specifications

Antigen ZNF683
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description zinc finger protein 683
Gene ZNF683
Gene Accession No. NM_173574.1
Gene Alias MGC33414
Host Species Mouse
Immunogen ZNF683 (NP_775845.1, 1 a.a. ∼ 509 a.a) full-length human protein
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 257101
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.