Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF683 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554902 Supplier Invitrogen™ Supplier No. PA554902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLLMMVNEL GHPSARWETL LPYPGAFQAS GQALPSQARN PGAGAAPTDS PGLERGGMAS PAKRVPLSSQ TGTAALPYPL KKKNGKILYE C Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 31%.

This gene, ZNF683, which is located on chromosome 1, is predicted to encode a zinc finger protein, currently with unknown fucntion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF683
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF683
Gene Accession No. Q8IZ20
Gene Alias Hobit; homolog of Blimp-1 in T cells; Homolog of Blimp-1 in T-cell; hypothetical protein MGC33414; MGC33414; RP11-569G9.6; Tissue-resident T-cell transcription regulator protein ZNF683; Zinc finger protein 683; ZNF683
Gene Symbols ZNF683
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF683. Recombinant protein control fragment (Product #RP-93613).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257101
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.