Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF684 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540984 Supplier Invitrogen™ Supplier No. PA540984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ANPHESSPES DYPLVDEPGK HRESKDNFLK SVLLTFNKIL TMERIHHYNM Sequence homology: Human: 100%; Pig: 79%.

ZNF684 contains 1 KRAB domain. The function of the ZNF684 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF684
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF684
Gene Accession No. Q5T5D7
Gene Alias hypothetical protein MGC27466; zinc finger protein 684; ZNF684
Gene Symbols ZNF684
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF684 (aa 72-121).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127396
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.