Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF687 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554347 Supplier Invitrogen™ Supplier No. PA554347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EETAGKGAGG ALLTPKTEPE ELAVSQGGAA PATEESSSSS EEEEVPSSPE PPRPAKRPRR ELGSKGLKGG Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

The zinc finger protein 687 (ZNF687) was initially identified as a translocation partner gene with RUNX1 in patients with acute myeloid leukemia (AML). Little is known of the function of the ZNF687 protein, but it has been shown to weakly interact with the Ring1/Rnf2 RING finger protein member of the Polycomb group of proteins, suggesting it may be involved in the chromatin-modifying complexes essential for embryonic development and stem cell renewal. Other evidence suggests that ZNF687 may be part of a transcriptional network that also includes ZNF592 and ZMYMD8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF687
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Znf687
Gene Accession No. Q8N1G0
Gene Alias KIAA1441; PDB6; RP11-126K1.3; Zinc finger protein 687; ZNF687
Gene Symbols Znf687
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF687. Recombinant protein control fragment (Product #RP-93581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.